missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dimethyl-1-phenyl-1H-pyrazole-4-sulfonyl chloride, ≥97%, Maybridge

Chemical Identifiers

CAS 59340-26-0
CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 97+%
Molecular Formula C11H11ClN2O2S
Formula Weight 270.74
MDL Number MFCD04968669
Chemical Name or Material 3, 5-Dimethyl-1-phenyl-1H-pyrazole-4-sulfonyl chloride
Assay Tech.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC48003CB SDS Fisher Scientific Company
CC48003CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC48003DA SDS Fisher Scientific Company
CC48003DA
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

59340-26-0
100.0
C11H11ClN2O2S
MFCD04968669
Tech.
80.0
97+%
270.74
3, 5-Dimethyl-1-phenyl-1H-pyrazole-4-sulfonyl chloride

Specifications

250mg
80.0
Amber Glass Bottle
C11H11ClN2O2S
MFCD04968669
Tech.
59340-26-0
100.0
97+%
270.74
3, 5-Dimethyl-1-phenyl-1H-pyrazole-4-sulfonyl chloride
Technical Grade

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.