Please login to your online account to display your discounted pricing

3,5-Dimethyl-1-phenyl-1H-pyrazole-4-carboxylic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3, 5-Dimethyl-1-phenyl-1H-pyrazole-4-carboxylic acid
CAS 61226-19-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H12N2O2
Formula Weight 216.24
MDL Number MFCD02656670
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00744DA

 
fisher scientific company
MO00744DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 216.24
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 3, 5-Dimethyl-1-phenyl-1H-pyrazole-4-carboxylic acid
Molecular Formula C12H12N2O2
MDL Number MFCD02656670
CAS 61226-19-5
Packaging Amber Glass Bottle
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.