Please login to your online account to display your discounted pricing

3,5-Dimethyl-1-hexyn-3-ol, 99%, Acros Organics

Chemical Identifiers

Chemical Name or Material 3, 5-Dimethyl-1-hexyn-3-ol
Assay Percent Range 99%
Molecular Formula C8H14O
Formula Weight 126.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC371555000

  SDS
thermo fisher scientific
AC371555000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC371551000

  SDS
thermo fisher scientific
AC371551000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 126.2
Quantity 500g
Chemical Name or Material 3, 5-Dimethyl-1-hexyn-3-ol
CAS 107-54-0
Packaging Glass bottle
Assay Percent Range 99%
Molecular Formula C8H14O