Please login to your online account to display your discounted pricing

3,5-Dimethyl-1-hexyn-3-ol, 99%, Acros Organics


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC371551000

 View more versions of this product

Catalog No. AC371551000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 126.2
Quantity 100g
Chemical Name or Material 3, 5-Dimethyl-1-hexyn-3-ol
Packaging Glass bottle
Assay Percent Range 99%
Molecular Formula C8H14O
CAS 107-54-0

Specifications

Chemical Name or Material 3, 5-Dimethyl-1-hexyn-3-ol
Assay Percent Range 99%
Molecular Formula C8H14O
Formula Weight 126.2