missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dimethoxybenzoyl Chloride 98.0+%, TCI America™

$87.92 - $245.58

Chemical Identifiers

CAS 17213-57-9
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.618
MDL Number MFCD00000676
InChI Key FTHPLWDYWAKYCY-UHFFFAOYSA-N
Synonym 3,5-dimethoxy benzoyl chloride, 3,5-dimethoxy-1-benzenecarbonyl chloride, 3,5-dimethoxy-benzoyl chloride, 3,5-dimethoxybenzoic acid chloride, 3,5-dimethoxybenzoyl chloride, acmc-1c560, benzoyl chloride, 3,5-dimethoxy, benzoyl chloride,3,5-dimethoxy
PubChem CID 87003
IUPAC Name 3,5-dimethoxybenzoyl chloride
SMILES COC1=CC(=CC(=C1)C(=O)Cl)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D26095G
View Documents
Tci America
D26095G
5g Each for $87.92
 
D260925G
View Documents
Tci America
D260925G
25g Each for $245.58
 
Specifications

Chemical Identifiers

17213-57-9
200.618
FTHPLWDYWAKYCY-UHFFFAOYSA-N
87003
COC1=CC(=CC(=C1)C(=O)Cl)OC
C9H9ClO3
MFCD00000676
3,5-dimethoxy benzoyl chloride, 3,5-dimethoxy-1-benzenecarbonyl chloride, 3,5-dimethoxy-benzoyl chloride, 3,5-dimethoxybenzoic acid chloride, 3,5-dimethoxybenzoyl chloride, acmc-1c560, benzoyl chloride, 3,5-dimethoxy, benzoyl chloride,3,5-dimethoxy
3,5-dimethoxybenzoyl chloride

Specifications

White-Yellow
17213-57-9
MFCD00000676
3,5-dimethoxy benzoyl chloride, 3,5-dimethoxy-1-benzenecarbonyl chloride, 3,5-dimethoxy-benzoyl chloride, 3,5-dimethoxybenzoic acid chloride, 3,5-dimethoxybenzoyl chloride, acmc-1c560, benzoyl chloride, 3,5-dimethoxy, benzoyl chloride,3,5-dimethoxy
COC1=CC(=CC(=C1)C(=O)Cl)OC
200.618
200.62
≥98.0% (GC,T)
5g
C9H9ClO3
3261
FTHPLWDYWAKYCY-UHFFFAOYSA-N
3,5-dimethoxybenzoyl chloride
87003
Crystal-Lump at 20°C
3,5-Dimethoxybenzoyl Chloride
Safety and Handling

Safety and Handling

TSCA : No