Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dimethoxybenzoyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 3,5-Dimethoxybenzoyl chloride
CAS 17213-57-9
Molecular Formula C9H9ClO3
Formula Weight 200.62
MDL Number MFCD00000676
UN Number UN3261
Beilstein 511839
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1474406

 
alfa aesar™
A1474406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1474414

 
alfa aesar™
A1474414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1474422

 
alfa aesar™
A1474422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 200.62
Chemical Name or Material 3,5-Dimethoxybenzoyl chloride
Assay 98%
Molecular Formula C9H9ClO3
Melting Point 42° to 44°C
Boiling Point 152° to 154°C (11mmHg)
MDL Number MFCD00000676
CAS 17213-57-9
UN Number UN3261
Beilstein 511839
Quantity 5g
Flash Point >110°C (230°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Applications
3,5-Dimethoxybenzoyl chloride was used to study the mechanism and kinetics of solvolysis of 3,4- and 3,5-dimethoxybenzoyl chlorides in various binary solvents. 3,5-Dimethoxybenzoyl chloride undergoes addition reaction with 4,4-dimethyl-2-pentyne in presence of AlCl3 via 1,2- methyl shift. It is used as a chemical and organic intermediate.

Solubility
It hydrolyzes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong bases.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.