missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dimethoxybenzoyl chloride, 98%

Manufacturer:  Alfa Aesar™ A1474422

 View more versions of this product

Catalog No. AAA1474422



Description

Description

Applications
3,5-Dimethoxybenzoyl chloride was used to study the mechanism and kinetics of solvolysis of 3,4- and 3,5-dimethoxybenzoyl chlorides in various binary solvents. 3,5-Dimethoxybenzoyl chloride undergoes addition reaction with 4,4-dimethyl-2-pentyne in presence of AlCl3 via 1,2- methyl shift. It is used as a chemical and organic intermediate.

Solubility
It hydrolyzes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong bases.
Specifications

Specifications

3,5-Dimethoxybenzoyl chloride
152°C to 154°C (11mmHg)
100g
C9H9ClO3
UN3261
benzoyl chloride, 3,5-dimethoxy, 3,5-dimethoxy benzoyl chloride, 3,5-dimethoxy-benzoyl chloride, acmc-1c560, benzoyl chloride,3,5-dimethoxy, 3,5-dimethoxybenzoic acid chloride, 3,5-dimethoxybenzoyl chloride, 3,5-dimethoxy-1-benzenecarbonyl chloride
COC1=CC(=CC(=C1)C(=O)Cl)OC
200.618
200.62
>110°C (230°F)
42°C to 44°C
17213-57-9
MFCD00000676
511839
FTHPLWDYWAKYCY-UHFFFAOYSA-N
3,5-dimethoxybenzoyl chloride
87003
98%

Chemical Identifiers

17213-57-9
200.618
FTHPLWDYWAKYCY-UHFFFAOYSA-N
87003
COC1=CC(=CC(=C1)C(=O)Cl)OC
C9H9ClO3
MFCD00000676
benzoyl chloride, 3,5-dimethoxy, 3,5-dimethoxy benzoyl chloride, 3,5-dimethoxy-benzoyl chloride, acmc-1c560, benzoyl chloride,3,5-dimethoxy, 3,5-dimethoxybenzoic acid chloride, 3,5-dimethoxybenzoyl chloride, 3,5-dimethoxy-1-benzenecarbonyl chloride
3,5-dimethoxybenzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

SDS Documents