missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,5-Dimethoxybenzoic acid, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 115461000
Specifications
3, 5-Dimethoxybenzoic acid | |
Glass bottle | |
182°C to 185°C | |
1132-21-4 | |
100.0 | |
C9H10O4 | |
10, 405 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol and ether | |
COC1=CC(=CC(=C1)C(=O)O)OC | |
182.17 | |
182.17 | |
99% |
Authentic | |
Beige-Pink to White | |
100g | |
98.5 | |
98.5% min. (HPLC) | |
(CH3O)2C6H3CO2H | |
3,5-dimethoxy benzoic acid, 3,5-dimethoxy-benzoic acid, 3,5-dimethoxybenzoicacid, 38my78eckk, acmc-1c4zl, benzoic acid, 3,5-dimethoxy, benzoicacid, 3,5-dimethoxy, ksc176m0p, pubchem3121, unii-38my78eckk | |
IWPZKOJSYQZABD-UHFFFAOYSA-N | |
3,5-dimethoxybenzoic acid | |
14332 | |
Crystalline Powder |
Chemical Identifiers
1132-21-4 | |
182.17 | |
3,5-dimethoxy benzoic acid, 3,5-dimethoxy-benzoic acid, 3,5-dimethoxybenzoicacid, 38my78eckk, acmc-1c4zl, benzoic acid, 3,5-dimethoxy, benzoicacid, 3,5-dimethoxy, ksc176m0p, pubchem3121, unii-38my78eckk | |
3,5-dimethoxybenzoic acid |
C9H10O4 | |
IWPZKOJSYQZABD-UHFFFAOYSA-N | |
14332 | |
COC1=CC(=CC(=C1)C(=O)O)OC |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
May cause respiratory irritation.
GHS P Statement
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective glov
GHS Signal Word: Warning
EINECSNumber : 214-473-8