Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dimethoxybenzamide, 98%

$71.27 - $71.27

Chemical Identifiers

Chemical Name or Material 3,5-Dimethoxybenzamide
CAS 17213-58-0
Molecular Formula C9H11NO3
Formula Weight 181.19
MDL Number MFCD00007987
Beilstein 2641361
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2481714

 
alfa aesar™
B2481714
25g Each for $71.27
Description & Specifications

Specifications

Formula Weight 181.19
Chemical Name or Material 3,5-Dimethoxybenzamide
Assay 98%
Molecular Formula C9H11NO3
Melting Point 142° to 146°C
MDL Number MFCD00007987
CAS 17213-58-0
Beilstein 2641361
Quantity 25g