Learn More
3,5-Dimethoxyaniline, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 115361000
Specifications
3, 5-Dimethoxyaniline | |
Authentic | |
178.0°C (20.0 mmHg) | |
53.0°C to 55.0°C | |
98% | |
97.5 | |
97.5% min. (GC) | |
(CH3O)2C6H3NH2 | |
3,5-dimethoxy aniline, 3,5-dimethoxy-phenylamine, 3,5-dimethoxyanaline, 3,5-dimethoxybenzenamine, 3,5-dimethoxybenzeneamine, 3,5-dimethoxyphenylamine, 5-aminoresorcinol dimethyl ether, 5-aminoresorcinol dimethylether, benzenamine, 3,5-dimethoxy, pubchem4423 | |
WNRGWPVJGDABME-UHFFFAOYSA-N | |
3,5-dimethoxyaniline | |
66301 | |
Liquid |
135°C | |
Glass bottle | |
Brown to Colorless | |
100g | |
10272-07-8 | |
100.0 | |
C8H11NO2 | |
MFCD00008392 | |
Solubility in water: insoluble | |
COC1=CC(=CC(=C1)N)OC | |
153.18 | |
153.18 | |
98% |
Chemical Identifiers
10272-07-8 | |
153.18 | |
WNRGWPVJGDABME-UHFFFAOYSA-N | |
66301 | |
COC1=CC(=CC(=C1)N)OC |
C8H11NO2 | |
MFCD00008392 | |
3,5-dimethoxy aniline, 3,5-dimethoxy-phenylamine, 3,5-dimethoxyanaline, 3,5-dimethoxybenzenamine, 3,5-dimethoxybenzeneamine, 3,5-dimethoxyphenylamine, 5-aminoresorcinol dimethyl ether, 5-aminoresorcinol dimethylether, benzenamine, 3,5-dimethoxy, pubchem4423 | |
3,5-dimethoxyaniline |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 233-616-5
TSCA : TSCA