missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dimethoxyaniline, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 115360250

 View more versions of this product

Catalog No. AC115360250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3, 5-Dimethoxyaniline
Authentic
178.0°C (20.0 mmHg)
53.0°C to 55.0°C
98%
97.5
97.5% min. (GC)
(CH3O)2C6H3NH2
3,5-dimethoxy aniline, 3,5-dimethoxy-phenylamine, 3,5-dimethoxyanaline, 3,5-dimethoxybenzenamine, 3,5-dimethoxybenzeneamine, 3,5-dimethoxyphenylamine, 5-aminoresorcinol dimethyl ether, 5-aminoresorcinol dimethylether, benzenamine, 3,5-dimethoxy, pubchem4423
WNRGWPVJGDABME-UHFFFAOYSA-N
3,5-dimethoxyaniline
66301
Liquid
135°C
Glass bottle
Brown to Colorless
25g
10272-07-8
100.0
C8H11NO2
MFCD00008392
Solubility in water: insoluble
COC1=CC(=CC(=C1)N)OC
153.18
153.18
98%

Chemical Identifiers

10272-07-8
153.18
WNRGWPVJGDABME-UHFFFAOYSA-N
66301
COC1=CC(=CC(=C1)N)OC
C8H11NO2
MFCD00008392
3,5-dimethoxy aniline, 3,5-dimethoxy-phenylamine, 3,5-dimethoxyanaline, 3,5-dimethoxybenzenamine, 3,5-dimethoxybenzeneamine, 3,5-dimethoxyphenylamine, 5-aminoresorcinol dimethyl ether, 5-aminoresorcinol dimethylether, benzenamine, 3,5-dimethoxy, pubchem4423
3,5-dimethoxyaniline
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 233-616-5

TSCA : TSCA

SDS Documents