missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ 3,5-Dimethoxyaniline, 98%
Manufacturer: Alfa Aesar™ A1507318
Specifications
3,5-Dimethoxyaniline | |
Characteristic | |
53°C to 57°C | |
10272-07-8 | |
MFCD00008392 | |
638013 | |
WNRGWPVJGDABME-UHFFFAOYSA-N | |
3,5-dimethoxyaniline | |
66301 | |
98% |
>110°C (230°F) | |
178°C (20mmHg) | |
50g | |
C8H11NO2 | |
UN2811 | |
benzenamine, 3,5-dimethoxy, 3,5-dimethoxybenzeneamine, 5-aminoresorcinol dimethyl ether, 3,5-dimethoxy-phenylamine, 3,5-dimethoxyanaline, 3,5-dimethoxy aniline, 5-aminoresorcinol dimethylether, 3,5-dimethoxybenzenamine, 3,5-dimethoxyphenylamine, pubchem4423 | |
COC1=CC(=CC(=C1)N)OC | |
153.181 | |
153.18 |
Chemical Identifiers
10272-07-8 | |
153.181 | |
WNRGWPVJGDABME-UHFFFAOYSA-N | |
66301 | |
COC1=CC(=CC(=C1)N)OC |
C8H11NO2 | |
MFCD00008392 | |
benzenamine, 3,5-dimethoxy, 3,5-dimethoxybenzeneamine, 5-aminoresorcinol dimethyl ether, 3,5-dimethoxy-phenylamine, 3,5-dimethoxyanaline, 3,5-dimethoxy aniline, 5-aminoresorcinol dimethylether, 3,5-dimethoxybenzenamine, 3,5-dimethoxyphenylamine, pubchem4423 | |
3,5-dimethoxyaniline |
Safety and Handling
GHS H Statement
H311-H373-H302-H332
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
P280h-P309-P310-P302+P352
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Danger
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : TSCA