Learn More
3',5'-Dimethoxyacetophenone, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2522403
Description
3',5'-Dimethoxyacetophenone is used as starting reagent in the synthesis of 5,7-dimethoxy-3-methylindazole1. It was used in the synthesis of (R)-(+)-1-(3,5-dimethoxyphenyl)ethyl acetate-ether-180.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3′,5′-Dimethoxyacetophenone is used as starting reagent in the synthesis of 5,7-dimethoxy-3-methylindazole1. It was used in the synthesis of (R)-(+)-1-(3,5-dimethoxyphenyl)ethyl acetate-ether-180.
Solubility
Insoluble in water.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
3',5'-Dimethoxyacetophenone | |
39°C to 41°C | |
>110°C (230°F) | |
1.543 | |
1 g | |
3',5'-dimethoxyacetophenone, 1-3,5-dimethoxyphenyl ethanone, 3,5-dimethoxyacetophenone, 1-3,5-dimethoxyphenyl ethan-1-one, ethanone, 1-3,5-dimethoxyphenyl, 1-acetyl-3,5-dimethoxybenzene, acetophenone, 3',5'-dimethoxy-6ci,7ci, acmc-20anci, pubchem3391, opera_id_1889 | |
YJKHOUIVWKQRSL-UHFFFAOYSA-N | |
1-(3,5-dimethoxyphenyl)ethanone | |
95997 | |
97% |
39151-19-4 | |
290°C to 291°C | |
C10H12O3 | |
MFCD00008739 | |
2045586 | |
Insoluble in water. | |
COC1=CC(=CC(OC)=C1)C(C)=O | |
180.20 | |
180.2 |
Chemical Identifiers
39151-19-4 | |
180.20 | |
YJKHOUIVWKQRSL-UHFFFAOYSA-N | |
95997 | |
COC1=CC(=CC(OC)=C1)C(C)=O |
C10H12O3 | |
MFCD00008739 | |
3',5'-dimethoxyacetophenone, 1-3,5-dimethoxyphenyl ethanone, 3,5-dimethoxyacetophenone, 1-3,5-dimethoxyphenyl ethan-1-one, ethanone, 1-3,5-dimethoxyphenyl, 1-acetyl-3,5-dimethoxybenzene, acetophenone, 3',5'-dimethoxy-6ci,7ci, acmc-20anci, pubchem3391, opera_id_1889 | |
1-(3,5-dimethoxyphenyl)ethanone |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 254-322-3
RTECSNumber : KM5775717
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only