Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dimethoxy-4-hydroxycinnamic acid, Ultrapure MALDI Matrix


For use as a matrix in MALDI mass spectrometry analysis

Quantity 5 x 10mg

Manufacturer: alfa aesar™  J67683EXK

 View more versions of this product

Catalog No. AAJ67683EXK

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 224.21
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 530-59-6
Chemical Name or Material 3,5-Dimethoxy-4-hydroxycinnamic acid
Synonym Sinapinic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00004401
Beilstein 2130006
Color Pale yellow
Physical Form Powder
Health Hazard 1 Warning
Quantity 5 x 10mg
Melting Point ≈195°C (dec.)
Name Note Ultrapure MALDI Matrix

Specifications

Chemical Name or Material 3,5-Dimethoxy-4-hydroxycinnamic acid
CAS 530-59-6
Name Note Ultrapure MALDI Matrix
Formula Weight 224.21
MDL Number MFCD00004401
Beilstein 2130006
Synonym Sinapinic acid
Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents