Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dimethoxy-4-hydroxycinnamic acid, Ultrapure MALDI Matrix

For use as a matrix in MALDI mass spectrometry analysis

Chemical Identifiers

Chemical Name or Material 3,5-Dimethoxy-4-hydroxycinnamic acid
CAS 530-59-6
Name Note Ultrapure MALDI Matrix
Formula Weight 224.21
MDL Number MFCD00004401
Beilstein 2130006
Synonym Sinapinic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67683EXK

  SDS
alfa aesar™
J67683EXK
5 x 10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 224.21
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 530-59-6
Chemical Name or Material 3,5-Dimethoxy-4-hydroxycinnamic acid
Synonym Sinapinic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00004401
Beilstein 2130006
Color Pale yellow
Physical Form Powder
Health Hazard 1 Warning
Quantity 5 x 10mg
Melting Point ≈195°C (dec.)
Name Note Ultrapure MALDI Matrix
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.