Learn More
3,5-Dimethoxy-4-hydroxycinnamic acid, 98%, predominantly trans isomer, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 115550050
Specifications
3, 5-Dimethoxy-4-hydroxycinnamic acid | |
Glass bottle | |
203.0°C to 205.0°C | |
530-59-6 | |
100.0 | |
C11H12O5 | |
10,508 | |
Solubility in water: insoluble in water. Other solubilities: soluble in methanol,ethanol and acetone | |
COC1=CC(=CC(=C1O)OC)C=CC(=O)O | |
224.21 | |
CHEBI:15714 | |
Crystalline Powder |
Authentic | |
Beige-Yellow to Yellow | |
5g | |
97.5 | |
98% | |
HOC6H2(OCH3)2CH=CHCO2H | |
2e-3-4-hydroxy-3,5-dimethoxyphenyl prop-2-enoic acid, 3,5-dimethoxy-4-hydroxycinnamic acid, 4-hydroxy-3,5-dimethoxycinnamic acid, sinapate, sinapic acid, sinapinate, sinapinic acid, synapoic acid, trans-sinapic acid, unii-p0i60993ec | |
PCMORTLOPMLEFB-ONEGZZNKSA-N | |
(E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid | |
637775 | |
224.21 | |
98% |
Chemical Identifiers
530-59-6 | |
224.21 | |
2e-3-4-hydroxy-3,5-dimethoxyphenyl prop-2-enoic acid, 3,5-dimethoxy-4-hydroxycinnamic acid, 4-hydroxy-3,5-dimethoxycinnamic acid, sinapate, sinapic acid, sinapinate, sinapinic acid, synapoic acid, trans-sinapic acid, unii-p0i60993ec | |
CHEBI:15714 | |
COC1=CC(=CC(=C1O)OC)C=CC(=O)O |
C11H12O5 | |
PCMORTLOPMLEFB-ONEGZZNKSA-N | |
637775 | |
(E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 208-487-3