Please login to your online account to display your discounted pricing

3,5-Dimethoxy-4-hydroxybenzaldehyde, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC132881000

 View more versions of this product

Catalog No. AC132881000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 182.18
Quantity 100g
Color Light yellow-green to brown
Density 1.013g/mL
Assay 97.5 to 102.5%
Melting Point 110° to 113°C
Merck Index 15, 9145
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C9H10O4
MDL Number MFCD00006943
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material 3, 5-Dimethoxy-4-hydroxybenzaldehyde
Packaging Glass bottle
CAS 134-96-3
Synonym 4-Hydroxy-3, 5-dimethoxybenzaldehyde
CAS Min % 97.5

Specifications

Chemical Name or Material 3, 5-Dimethoxy-4-hydroxybenzaldehyde
CAS 134-96-3
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5 to 102.5%
Assay Percent Range 98%
Molecular Formula C9H10O4
Formula Weight 182.18
MDL Number MFCD00006943
Synonym 4-Hydroxy-3, 5-dimethoxybenzaldehyde
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents