missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3',5'-Dimethoxy-4'-hydroxyacetophenone, 97%, ACROS Organics™

$56.10 - $172.82

Chemical Identifiers

CAS 2478-38-8
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H12O4
Linear Formula HOC6H2(OCH3)2COCH3
Formula Weight 196.2
MDL Number MFCD00008748
Synonym Acetosyringone
Chemical Name or Material 3';, 5';-Dimethoxy-4';-hydroxyacetophenone, 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC115540010 SDS Fisher Scientific Acros
115540010
Glass bottle 1g Each for $56.10
AC115540050 SDS Fisher Scientific Acros
115540050
Glass bottle 5g Each for $172.82
AC115540250 SDS Thermo Fisher Scientific
AC115540250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

2478-38-8
100.0
C10H12O4
196.2
Acetosyringone
≥96.0%
96.0
97%
HOC6H2(OCH3)2COCH3
MFCD00008748
3';, 5';-Dimethoxy-4';-hydroxyacetophenone, 97%

Specifications

2478-38-8
100.0
Glass bottle
97%
123° to 128°C
Beige to light brown
MFCD00008748
Acetosyringone
≥96.0%
96.0
Authentic
Crystalline Powder
C10H12O4
HOC6H2(OCH3)2COCH3
196.2
1g
3';, 5';-Dimethoxy-4';-hydroxyacetophenone, 97%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 219-610-5

RTECSNumber : AM8440000

TSCA : TSCA