Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Diisopropylsalicylic acid, 98%

$86.35 - $270.92

Chemical Identifiers

Chemical Name or Material 3,5-Diisopropylsalicylic acid
CAS 2215-21-6
Molecular Formula C13H18O3
Formula Weight 222.28
MDL Number MFCD00008884
Beilstein 2505176
Synonym 2-Hydroxy-3,5-diisopropylbenzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1584514

 
alfa aesar™
A1584514
25g Each for $86.35

AAA1584522

 
alfa aesar™
A1584522
100g Each for $270.92
Description & Specifications

Specifications

Formula Weight 222.28
Chemical Name or Material 3,5-Diisopropylsalicylic acid
TSCA TSCA
Melting Point 115° to 119°C
Assay 98%
Molecular Formula C13H18O3
Quantity 25g
MDL Number MFCD00008884
CAS 2215-21-6
Beilstein 2505176
Synonym 2-Hydroxy-3,5-diisopropylbenzoic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
3,5-Diisopropylsalicylic acid is used to prepare 2,4-Diisopropyl-phenol at 200°C. It is also used as a starting reagent in the synthesis of Zn(II) and Cd(II) carboxylate complexes. It plays the role of OH-inactivating ligand in its complexes with copper(II)2.

Solubility
Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.