Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Diiodosalicylic acid lithium salt hydrate, 95%

$87.43 - $1,074.42

Chemical Identifiers

Chemical Name or Material 3,5-Diiodosalicylic acid lithium salt hydrate
CAS 653-14-5
Molecular Formula C7H3I2LiO3·xH2O
Formula Weight 413.86 (395.85 Anhydrous)
MDL Number MFCD00156142
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1578406

 
alfa aesar™
A1578406
5g Each for $87.43

AAA1578414

 
alfa aesar™
A1578414
25g Each for $335.57

AAA1578422

 
alfa aesar™
A1578422
100g Each for $1,074.42
Description & Specifications

Specifications

Formula Weight 413.86 (395.85 Anhydrous)
CAS 653-14-5
Chemical Name or Material 3,5-Diiodosalicylic acid lithium salt hydrate
TSCA TSCA
Assay 95%
Molecular Formula C7H3I2LiO3·xH2O
Quantity 5g
MDL Number MFCD00156142
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage −30° to −10°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

−30° to −10°C