Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Diiodo-L-tyrosine hydrate, 98% (dry wt.), may contain up to ≈10% water

$380.20 - $380.20

Chemical Identifiers

Chemical Name or Material 3,5-Diiodo-L-tyrosine hydrate
CAS 18835-59-1
Molecular Formula C9H9I2NO3·xH2O
Formula Weight 432.98 (Anhydrous)
MDL Number MFCD00150275
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1070222

 
alfa aesar™
A1070222
100g Each for $380.20
Description & Specifications

Specifications

Formula Weight 432.98 (Anhydrous)
Chemical Name or Material 3,5-Diiodo-L-tyrosine hydrate
TSCA TSCA
Assay 98%
Assay Percent Notes (dry wt.)
Molecular Formula C9H9I2NO3·xH2O
MDL Number MFCD00150275
CAS 18835-59-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 185°C (dec.)
Quantity 100g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Concentration May contain up to ≈10% water
Recommended Storage 2°C to 8°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2°C to 8°C