Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Diiodo-L-tyrosine, 95%

Chemical Identifiers

Chemical Name or Material 3,5-Diiodo-L-tyrosine
CAS 300-39-0
Formula Weight 432.98
MDL Number MFCD00016542
Beilstein 2218691
Synonym H-Tyr(3,5-Di-I)-OH; (S)-2-Amino-3-(4-hydroxy-3,5-diiodophenyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6542014

  SDS
alfa aesar™
H6542014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6542022

  SDS
alfa aesar™
H6542022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 432.98
Chemical Name or Material 3,5-Diiodo-L-tyrosine
Assay 95%
Physical Form Solid
MDL Number MFCD00016542
Beilstein 2218691
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Color Pale yellow
Quantity 25g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
CAS 300-39-0
Synonym H-Tyr(3,5-Di-I)-OH; (S)-2-Amino-3-(4-hydroxy-3,5-diiodophenyl)propionic acid
Sensitivity Light Sensitive
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.