Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Diiodo-4-pyridone-1-acetic acid, 99%

$38.95 - $314.01

Chemical Identifiers

Chemical Name or Material 3,5-Diiodo-4-pyridone-1-acetic acid
CAS 101-29-1
Molecular Formula C7H5I2NO3
Formula Weight 404.93
MDL Number MFCD00006185
Beilstein 163586
Synonym 3,5-Diiodo-4-oxo-1(4H)-pyridineacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2564603

  SDS
alfa aesar™
B2564603
1g Each for $38.95

AAB2564609

  SDS
alfa aesar™
B2564609
10g Each for $314.01
Description & Specifications

Specifications

Formula Weight 404.93
Chemical Name or Material 3,5-Diiodo-4-pyridone-1-acetic acid
Melting Point 244°C (dec.)
Assay 99%
Molecular Formula C7H5I2NO3
Quantity 1g
MDL Number MFCD00006185
CAS 101-29-1
Beilstein 163586
Synonym 3,5-Diiodo-4-oxo-1(4H)-pyridineacetic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.