Please login to your online account to display your discounted pricing

3,5-Dihydroxybenzyl alcohol, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 5-Dihydroxybenzyl alcohol
CAS 29654-55-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H8O3
Linear Formula (HO)C6H3CHOH
Formula Weight 140.14
MDL Number MFCD00016867
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC344060250

  SDS
thermo fisher scientific
AC344060250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC344060050

  SDS
fisher scientific acros
344060050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 140.14
Quantity 25g
Color Off-white to beige-pink or gray
Melting Point 181° to 186°C
CAS Max % 100.0
Chemical Name or Material 3, 5-Dihydroxybenzyl alcohol
Assay Percent Range 98%
Molecular Formula C7H8O3
CAS 29654-55-5
MDL Number MFCD00016867
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Linear Formula (HO)C6H3CHOH
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.