Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dihydroxybenzyl alcohol, 98%

$46.26 - $552.99

Chemical Identifiers

Chemical Name or Material 3,5-Dihydroxybenzyl alcohol
CAS 29654-55-5
Molecular Formula C7H8O3
Formula Weight 140.14
MDL Number MFCD00016867
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2524803

  SDS
alfa aesar™
B2524803
1g Each for $46.26

AAB2524814

  SDS
alfa aesar™
B2524814
25g Each for $552.99
Description & Specifications

Specifications

Formula Weight 140.14
Chemical Name or Material 3,5-Dihydroxybenzyl alcohol
Assay 98%
Molecular Formula C7H8O3
MDL Number MFCD00016867
CAS 29654-55-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 182° to 186°C (dec.)
Quantity 1g

Applications
3,5-Dihydroxybenzyl alcohol may be used as monomer for the synthesis of a series of monodisperse dendritic polyether macromolecules. It may be employed in the synthesis of dimethylsilyl linked dihydroxybenzyl alcohol based dendrimers.

Solubility
Soluble in DMSO, methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.