missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,5-Dihydroxybenzyl alcohol, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals B2524803
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications3,5-Dihydroxybenzyl alcohol may be used as monomer for the synthesis of a series of monodisperse dendritic polyether macromolecules. It may be employed in the synthesis of dimethylsilyl linked dihydroxybenzyl alcohol based dendrimers.
Solubility
Soluble in DMSO, methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Specifications
3,5-Dihydroxybenzyl alcohol | |
182°C to 186°C (decomposition) | |
MFCD00016867 | |
3,5-dihydroxybenzyl alcohol, 5-hydroxymethyl-1,3-benzenediol, 5-hydroxymethyl benzene-1,3-diol, 5-hydroxymethyl resorcinol, benzyl alcohol, 3,5-dihydroxy, 1,3-benzenediol, 5-hydroxymethyl, 3,5-dihydroxybenzylalcohol, 3,5-dihydroxy benzyl alcohol, pubchem7418, acmc-1cjog | |
NGYYFWGABVVEPL-UHFFFAOYSA-N | |
5-(hydroxymethyl)benzene-1,3-diol | |
34661 | |
98% |
29654-55-5 | |
C7H8O3 | |
1 g | |
Soluble in DMSO,methanol. | |
C1=C(C=C(C=C1O)O)CO | |
140.138 | |
140.14 |
Chemical Identifiers
29654-55-5 | |
140.138 | |
NGYYFWGABVVEPL-UHFFFAOYSA-N | |
34661 | |
C1=C(C=C(C=C1O)O)CO |
C7H8O3 | |
MFCD00016867 | |
3,5-dihydroxybenzyl alcohol, 5-hydroxymethyl-1,3-benzenediol, 5-hydroxymethyl benzene-1,3-diol, 5-hydroxymethyl resorcinol, benzyl alcohol, 3,5-dihydroxy, 1,3-benzenediol, 5-hydroxymethyl, 3,5-dihydroxybenzylalcohol, 3,5-dihydroxy benzyl alcohol, pubchem7418, acmc-1cjog | |
5-(hydroxymethyl)benzene-1,3-diol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 249-751-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only