missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dihydroxybenzoic acid, 98%

Manufacturer:  Alfa Aesar™ A1166422

 View more versions of this product

Catalog No. AAA1166422



Description

Description

Applications
3,5-Dihydroxybenzoic acid is used in organic synthesis and as intermediate for the pharmaceutical.

Solubility
Soluble in 50 mg/mL, 95% Ethanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

~237°C (decomposition)
3,5-Dihydroxybenzoic acid
98%
MFCD00002512
alpha-Resorcylic acid
C1=C(C=C(C=C1O)O)C(=O)O
154.121
CHEBI:39912
100g
99-10-5
C7H6O4
2207864
UYEMGAFJOZZIFP-UHFFFAOYSA-N
3,5-dihydroxybenzoic acid
7424
154.12

Chemical Identifiers

99-10-5
154.121
UYEMGAFJOZZIFP-UHFFFAOYSA-N
7424
3,5-dihydroxybenzoic acid
C7H6O4
MFCD00002512
alpha-Resorcylic acid
CHEBI:39912
C1=C(C=C(C=C1O)O)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : VH3708000

TSCA : TSCA

SDS Documents