missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dihydroxybenzoic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC114900025

 View more versions of this product

Catalog No. AC114900025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3,5-Dihydroxybenzoic acid
Authentic
235.0°C to 240.0°C
97%
96.0
96% min. (HPLC)
(HO)2C6H3CO2H
10,404
Solubility in water: 84g/L (20°C). Other solubilities: soluble in ethanol and ether
C1=C(C=C(C=C1O)O)C(=O)O
154.12
CHEBI:39912
Powder
>200°C
White to Beige
2.5kg
99-10-5
100.0
C7H6O4
MFCD00002512
.alpha.-resorcylic acid, 3,5-dhba, 3,5-dihydroxy benzoic acid, 3,5-dihydroxy-benzoic acid, 3,5-dihydroxybenzoicacid, 5-carboxyresorcinol, a-resorcylic acid, alpha-resorcylic acid, benzoic acid, 3,5-dihydroxy, unii-2wc5lmo6l1
UYEMGAFJOZZIFP-UHFFFAOYSA-N
3,5-dihydroxybenzoic acid
7424
154.12
97%

Chemical Identifiers

99-10-5
154.12
UYEMGAFJOZZIFP-UHFFFAOYSA-N
7424
3,5-dihydroxybenzoic acid
C7H6O4
MFCD00002512
.alpha.-resorcylic acid, 3,5-dhba, 3,5-dihydroxy benzoic acid, 3,5-dihydroxy-benzoic acid, 3,5-dihydroxybenzoicacid, 5-carboxyresorcinol, a-resorcylic acid, alpha-resorcylic acid, benzoic acid, 3,5-dihydroxy, unii-2wc5lmo6l1
CHEBI:39912
C1=C(C=C(C=C1O)O)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-730-7

RTECSNumber : VH3708000

TSCA : TSCA

SDS Documents