missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dihydroxybenzaldehyde, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 425650250

Catalog No. AC425650250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3, 5-Dihydroxybenzaldehyde
97.5
Authentic
Glass bottle
(HO)2C6H3CHO
25g
HAQLHRYUDBKTJG-UHFFFAOYSA-N
3,5-dihydroxybenzaldehyde
94365
138.12
26153-38-8
100.0
97.5% min. (HPLC)
C7H6O3
MFCD00016611
benzaldehyde, 3,5-dihydroxy, 3,5-dihydroxy benzaldehyde, 5-formylresorcinol, 5-formylbenzene-1,3-diol, pubchem8249, alpha-resorcylic aldehyde, acmc-1cg7h, 3,5-dihydoxybenzal dehyde, 3,5-dihydroxy-benzaldehyde, .alpha.-resorcylic aldehyde
OC1=CC(C=O)=CC(O)=C1
138.12
CHEBI:50204
98%
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye prote

GHS Signal Word: Warning

EINECSNumber : 247-479-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only