missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dihydroxybenzaldehyde, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1533806

 View more versions of this product

Catalog No. AAA1533806



Description

Description

Applications
3,5-Dihydroxybenzaldehyde is used as a building block in the synthesis of more complex structures. It is also used in the synthesis of terbutaline (T109750), which is an important bronchodilator.

Solubility
Soluble in ethanol and diethyl ether.

Notes
Air sensitive. Store in cool place. Incompatible with strong acids and strong bases.
Specifications

Specifications

3,5-Dihydroxybenzaldehyde
5g
98%
MFCD00016611
benzaldehyde, 3,5-dihydroxy, 3,5-dihydroxy benzaldehyde, 5-formylresorcinol, 5-formylbenzene-1,3-diol, pubchem8249, alpha-resorcylic aldehyde, acmc-1cg7h, 3,5-dihydoxybenzal dehyde, 3,5-dihydroxy-benzaldehyde, .alpha.-resorcylic aldehyde
C1=C(C=C(C=C1O)O)C=O
138.122
CHEBI:50204
98%
158°C to 161°C
26153-38-8
C7H6O3
1930147
HAQLHRYUDBKTJG-UHFFFAOYSA-N
3,5-dihydroxybenzaldehyde
94365
138.12

Chemical Identifiers

26153-38-8
138.122
HAQLHRYUDBKTJG-UHFFFAOYSA-N
94365
3,5-dihydroxybenzaldehyde
C7H6O3
MFCD00016611
benzaldehyde, 3,5-dihydroxy, 3,5-dihydroxy benzaldehyde, 5-formylresorcinol, 5-formylbenzene-1,3-diol, pubchem8249, alpha-resorcylic aldehyde, acmc-1cg7h, 3,5-dihydoxybenzal dehyde, 3,5-dihydroxy-benzaldehyde, .alpha.-resorcylic aldehyde
CHEBI:50204
C1=C(C=C(C=C1O)O)C=O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : Air sensitive

SDS Documents