Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Difluoropyridine, 98%

$54.69 - $54.69

Chemical Identifiers

Chemical Name or Material 3,5-Difluoropyridine
CAS 71902-33-5
Molecular Formula C5H3F2N
Formula Weight 115.08
MDL Number MFCD03412222
UN Number UN1993
Beilstein 7914363
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1953903

  SDS
alfa aesar™
L1953903*
1g Each for $54.69

AAL1953906

  SDS
alfa aesar™
L1953906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 115.08
Chemical Name or Material 3,5-Difluoropyridine
Assay 98%
Molecular Formula C5H3F2N
MDL Number MFCD03412222
CAS 71902-33-5
UN Number UN1993
Beilstein 7914363
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Flash Point 9°C (48°F)
Boiling Point 92° to 93°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.