Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Difluoropyridine-2-carboxamide, 97%

$87.28 - $332.49

Chemical Identifiers

Chemical Name or Material 3,5-Difluoropyridine-2-carboxamide
CAS 745784-03-6
Molecular Formula C6H4F2N2O
Formula Weight 158.11
MDL Number MFCD05664123
Synonym 3,5-Difluoropicolinamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2032303

 
alfa aesar™
L2032303
1g Each for $87.28

AAL2032306

 
alfa aesar™
L2032306
5g Each for $332.49
Description & Specifications

Specifications

Formula Weight 158.11
Chemical Name or Material 3,5-Difluoropyridine-2-carboxamide
Assay 97%
Molecular Formula C6H4F2N2O
MDL Number MFCD05664123
CAS 745784-03-6
Synonym 3,5-Difluoropicolinamide
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.