Please login to your online account to display your discounted pricing

Alfa Aesar™ 3',5'-Difluoropropiophenone, 97%

$174.78 - $693.11

Chemical Identifiers

Chemical Name or Material 3',5'-Difluoropropiophenone
CAS 135306-45-5
Molecular Formula C9H8F2O
Formula Weight 170.16
MDL Number MFCD00061142
Beilstein 7422838
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2015006

  SDS
alfa aesar™
B2015006
5g Each for $174.78

AAB2015014

  SDS
alfa aesar™
B2015014
25g Each for $693.11
Description & Specifications

Specifications

Formula Weight 170.16
Molecular Formula C9H8F2O
CAS 135306-45-5
Melting Point 25° to 27°C
Quantity 5g
Assay 97%
Boiling Point 82°C (10mmHg)
Chemical Name or Material 3',5'-Difluoropropiophenone
MDL Number MFCD00061142
Beilstein 7422838