missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Difluorophenylhydrazine hydrochloride, +97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC398830050

 View more versions of this product

Catalog No. AC398830050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

180.58
100.0
Glass bottle
C6H6F2N2·HCl
97+%
MFCD03094171
5g
97.0
134993-88-7
3, 5-Difluorophenylhydrazine hydrochloride
F2C6H3NHNH2·HCl

Chemical Identifiers

134993-88-7
100.0
C6H6F2N2·HCl
180.58
3, 5-Difluorophenylhydrazine hydrochloride
97.0
97+%
F2C6H3NHNH2·HCl
MFCD03094171

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning