Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Difluorophenylacetic acid, 99%

$876.93 - $876.93

Chemical Identifiers

Chemical Name or Material 3,5-Difluorophenylacetic acid
CAS 105184-38-1
Molecular Formula C8H6F2O2
Formula Weight 172.13
MDL Number MFCD00010316
Beilstein 3605064
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1948414

  SDS
alfa aesar™
A1948414
25g Each for $876.93
Description & Specifications

Specifications

Formula Weight 172.13
Health Hazard 1 Warning
Beilstein 3605064
Assay 99%
MDL Number MFCD00010316
Chemical Name or Material 3,5-Difluorophenylacetic acid
CAS 105184-38-1
Quantity 25g
Molecular Formula C8H6F2O2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 68° to 70°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.