Learn More
3,5-Difluorophenyl isocyanate, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 83594-83-6 |
---|---|
Molecular Formula | C7H3F2NO |
Molecular Weight (g/mol) | 155.10 |
MDL Number | MFCD02093693 |
InChI Key | WUCCJYOIAJTFFQ-UHFFFAOYSA-N |
Synonym | 3,5-difluoro-1-isocyanatobenzene, 3,5-difluorophenyl isocyanate, 3,5-difluorophenyl-isocyanate, 3,5-difluorophenylisocyanate, acmc-1bk6p, benzene, 1,3-difluoro-5-isocyanato, benzene, 1,3-difluoro-5-isocyanato-9ci, benzene,1,3-difluoro-5-isocyanato, ksc650q9d, pubchem4454 |
PubChem CID | 2779282 |
IUPAC Name | 1,3-difluoro-5-isocyanatobenzene |
SMILES | FC1=CC(=CC(F)=C1)N=C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC428230010
|
Thermo Fisher Scientific
AC428230010 |
1g | Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
83594-83-6 | |
155.10 | |
WUCCJYOIAJTFFQ-UHFFFAOYSA-N | |
2779282 | |
FC1=CC(=CC(F)=C1)N=C=O |
C7H3F2NO | |
MFCD02093693 | |
3,5-difluoro-1-isocyanatobenzene, 3,5-difluorophenyl isocyanate, 3,5-difluorophenyl-isocyanate, 3,5-difluorophenylisocyanate, acmc-1bk6p, benzene, 1,3-difluoro-5-isocyanato, benzene, 1,3-difluoro-5-isocyanato-9ci, benzene,1,3-difluoro-5-isocyanato, ksc650q9d, pubchem4454 | |
1,3-difluoro-5-isocyanatobenzene |
Specifications
83594-83-6 | |
100.0 | |
159.0°C | |
97% | |
F2C6H3NCO | |
Glass Bottle | |
1g | |
FC1=CC(=CC(F)=C1)N=C=O | |
155.10 | |
155.1 | |
3, 5-Difluorophenyl isocyanate |
96.0 | |
1.3100g/mL | |
54°C | |
C7H3F2NO | |
MFCD02093693 | |
3,5-difluoro-1-isocyanatobenzene, 3,5-difluorophenyl isocyanate, 3,5-difluorophenyl-isocyanate, 3,5-difluorophenylisocyanate, acmc-1bk6p, benzene, 1,3-difluoro-5-isocyanato, benzene, 1,3-difluoro-5-isocyanato-9ci, benzene,1,3-difluoro-5-isocyanato, ksc650q9d, pubchem4454 | |
WUCCJYOIAJTFFQ-UHFFFAOYSA-N | |
1,3-difluoro-5-isocyanatobenzene | |
2779282 | |
97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning