missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Difluorophenol, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC306610010

 View more versions of this product

Catalog No. AC306610010


 

Specifications Description & Specifications

Specifications

3, 5-Difluorophenol
51° to 55°C
Authentic
2713-34-0
100.0
Crystals
99%
F2C6H3OH
MFCD00002255
White to beige
1g
99%
98.5
Glass bottle
≥98.5%
C6H4F2O
130.09

Chemical Identifiers

2713-34-0
100.0
C6H4F2O
130.09
3, 5-Difluorophenol, 99%
98.5
99%
F2C6H3OH
MFCD00002255
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Flammable solid.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA