Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Difluoromandelic acid, 97%

$326.32 - $326.32

Chemical Identifiers

Chemical Name or Material 3,5-Difluoromandelic acid
CAS 132741-31-2
Molecular Formula C8H6F2O3
Formula Weight 188.13
MDL Number MFCD00061300
Synonym alpha-Hydroxy-3,5-difluorophenylacetic acid; 3,5-Difluoro-alpha-hydroxyphenylacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2242306

 
alfa aesar™
B2242306
5g Each for $326.32
Description & Specifications

Specifications

Formula Weight 188.13
Chemical Name or Material 3,5-Difluoromandelic acid
Assay 97%
Molecular Formula C8H6F2O3
MDL Number MFCD00061300
CAS 132741-31-2
Synonym alpha-Hydroxy-3,5-difluorophenylacetic acid; 3,5-Difluoro-alpha-hydroxyphenylacetic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 135° to 137°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.