Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Difluorobenzylmagnesium bromide, 0.25M in 2-MeTHF

Chemical Identifiers

Chemical Name or Material 3,5-Difluorobenzylmagnesium bromide
Formula Weight 231.32
MDL Number MFCD11113509
UN Number UN2924
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH54427AE

  SDS
alfa aesar™
H54427AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 231.32
UN Number UN2924
Sensitivity Air & Moisture Sensitive
Health Hazard 1 Danger
Concentration 0.25M in 2-MeTHF
Chemical Name or Material 3,5-Difluorobenzylmagnesium bromide
MDL Number MFCD11113509
DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Physical Form Liquid
Quantity 100mL
Health Hazard 2 GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
It is a green alternative to Tetrahydrofuran. is capable of producing high concentrations of bromo, benzyl and allyl Grignard reagents. 2-MeTHF quickly produces clean water phase splits without forming emulsions.

Solubility
Reacts with water.

Notes
Air & Moisture Sensitive. Store under inert dry gas.

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.