Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Difluorobenzyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 3,5-Difluorobenzyl chloride
CAS 220141-71-9
Molecular Formula C7H5ClF2
Formula Weight 162.57
MDL Number MFCD01090990
UN Number UN3265
Beilstein 7473788
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1964603

  SDS
alfa aesar™
L1964603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 162.57
Chemical Name or Material 3,5-Difluorobenzyl chloride
Health Hazard 1 Danger
Quantity 1g
Molecular Formula C7H5ClF2
CAS 220141-71-9
UN Number UN3265
MDL Number MFCD01090990
Assay 98%
Beilstein 7473788
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 82°C (22mmHg)
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.