Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Difluorobenzyl alcohol, 97%

$35.55 - $406.82

Chemical Identifiers

Chemical Name or Material 3,5-Difluorobenzyl alcohol
CAS 79538-20-8
Molecular Formula C7H6F2O
Formula Weight 144.12
MDL Number MFCD00010313
Beilstein 5499788
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1821303

  SDS
alfa aesar™
A1821303
1g Each for $35.55

AAA1821306

  SDS
alfa aesar™
A1821306
5g Each for $99.89

AAA1821314

  SDS
alfa aesar™
A1821314
25g Each for $406.82
Description & Specifications

Specifications

Formula Weight 144.12
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 98° to 99°C (9mmHg)
MDL Number MFCD00010313
Assay 97%
Health Hazard 1 Warning
Flash Point 94°C (201°F)
Molecular Formula C7H6F2O
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 79538-20-8
Beilstein 5499788
Chemical Name or Material 3,5-Difluorobenzyl alcohol
Quantity 1g

Applications
3,5-Difluorobenzyl alcohol is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.