Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Difluorobenzoic acid, 97%

$66.91 - $66.91

Chemical Identifiers

Chemical Name or Material 3,5-Difluorobenzoic acid
CAS 455-40-3
Molecular Formula C7H4F2O2
Formula Weight 158.11
MDL Number MFCD00010323
Beilstein 1940680
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1703306

  SDS
alfa aesar™
A1703306
5g Each for $66.91
Description & Specifications

Specifications

Formula Weight 158.11
MDL Number MFCD00010323
Assay 97%
Molecular Formula C7H4F2O2
Chemical Name or Material 3,5-Difluorobenzoic acid
Odor Odorless
Quantity 5g
CAS 455-40-3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 1 Warning
Beilstein 1940680
Melting Point 119° to 123°C
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.