missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Difluorobenzeneboronic acid neopentyl glycol ester, 98+%

Manufacturer:  Alfa Aesar™ L1726603

 View more versions of this product

Catalog No. AAL1726603



Specifications

Specifications

3,5-Difluorobenzeneboronic acid neopentyl glycol ester
1g
C11H13BF2O2
2-3,5-difluorophenyl-5,5-dimethyl-1,3,2-dioxaborinane, 3,5-difluorobenzeneboronic acid neopentyl glycol ester, 3,5-difluorophenylboronic acid neopentyl glycol ester, 3,5-difluorobenzeneboronic acid neopentyl glycol cyclic ester, 2-3,5-difluorophenyl-5,5-dimethyl-1,3,2-dioxaborane, 2-3,5-difluorophenyl-5,5-dimethyl-1,3,2-dioxaborine, 1,3,2-dioxaborinane, 2-3,5-difluorophenyl-5,5-dimethyl, 3,5-difluorobenzenebornoic acid neopentyl glycol cyclic ester, 2-3,5-difluorophenyl-5,5-dimethyldihydro-4h-1,3,2-dioxaborin, 3,5-difluorobenzenebornoic acid neopentyl glycol cyclic ester
B1(OCC(CO1)(C)C)C2=CC(=CC(=C2)F)F
226.03
226.03
43°C to 45°C
216393-57-6
MFCD01318139
OLHRAEPCGZMMTI-UHFFFAOYSA-N
2-(3,5-difluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
2774056
≥98%

Chemical Identifiers

216393-57-6
226.03
OLHRAEPCGZMMTI-UHFFFAOYSA-N
2774056
B1(OCC(CO1)(C)C)C2=CC(=CC(=C2)F)F
C11H13BF2O2
MFCD01318139
2-3,5-difluorophenyl-5,5-dimethyl-1,3,2-dioxaborinane, 3,5-difluorobenzeneboronic acid neopentyl glycol ester, 3,5-difluorophenylboronic acid neopentyl glycol ester, 3,5-difluorobenzeneboronic acid neopentyl glycol cyclic ester, 2-3,5-difluorophenyl-5,5-dimethyl-1,3,2-dioxaborane, 2-3,5-difluorophenyl-5,5-dimethyl-1,3,2-dioxaborine, 1,3,2-dioxaborinane, 2-3,5-difluorophenyl-5,5-dimethyl, 3,5-difluorobenzenebornoic acid neopentyl glycol cyclic ester, 2-3,5-difluorophenyl-5,5-dimethyldihydro-4h-1,3,2-dioxaborin, 3,5-difluorobenzenebornoic acid neopentyl glycol cyclic ester
2-(3,5-difluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
Documents