Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Difluorobenzeneboronic acid neopentyl glycol ester, 98+%


Quantity 5g

Manufacturer: alfa aesar™  L1726606

 View more versions of this product

Catalog No. AAL1726606

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 226.03
Chemical Name or Material 3,5-Difluorobenzeneboronic acid neopentyl glycol ester
Assay ≥98%
Molecular Formula C11H13BF2O2
MDL Number MFCD01318139
Synonym 2-(3,5-Difluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
CAS 216393-57-6
Melting Point 43° to 45°C
Quantity 5g

Specifications

Chemical Name or Material 3,5-Difluorobenzeneboronic acid neopentyl glycol ester
CAS 216393-57-6
Molecular Formula C11H13BF2O2
Formula Weight 226.03
MDL Number MFCD01318139
Synonym 2-(3,5-Difluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane