Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Difluorobenzamide, 97%

$35.72 - $128.07

Chemical Identifiers

Chemical Name or Material 3,5-Difluorobenzamide
CAS 132980-99-5
Molecular Formula C7H5F2NO
Formula Weight 157.12
MDL Number MFCD00061138
Beilstein 4177620
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1990403

  SDS
alfa aesar™
A1990403
1g Each for $35.72

AAA1990406

  SDS
alfa aesar™
A1990406
5g Each for $128.07
Description & Specifications

Specifications

Formula Weight 157.12
Chemical Name or Material 3,5-Difluorobenzamide
Assay 97%
Molecular Formula C7H5F2NO
MDL Number MFCD00061138
CAS 132980-99-5
Beilstein 4177620
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 155° to 157°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.