Learn More
3,5-Difluorobenzaldehyde, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 293470010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3, 5-Difluorobenzaldehyde | |
32085-88-4 | |
100.0 | |
56°C | |
98% min. (GC) | |
C7H4F2O | |
F2C6H3CHO | |
1g | |
benzaldehyde, 3,5-difluoro, 3,5-difluoro-benzaldehyde, pubchem1414, 3,5difluorobenzaldehyde, acmc-1aewe, 3,5-diflourobenzaldehyde, 3-5-difluorobenzaldehyde, 3, 5-difluorobenzaldehyde, ksc224o7p, 3,5-difluorophenyl methanone | |
C1=C(C=C(C=C1F)F)C=O | |
142.1 | |
142.1 |
98+% | |
98.0 | |
1.2960g/mL | |
Authentic | |
Glass bottle | |
1.4930 to 1.4950 | |
MFCD00010329 | |
1.296 | |
ASOFZHSTJHGQDT-UHFFFAOYSA-N | |
3,5-difluorobenzaldehyde | |
588160 | |
98+% |
Chemical Identifiers
32085-88-4 | |
142.1 | |
ASOFZHSTJHGQDT-UHFFFAOYSA-N | |
588160 | |
C1=C(C=C(C=C1F)F)C=O |
C7H4F2O | |
MFCD00010329 | |
benzaldehyde, 3,5-difluoro, 3,5-difluoro-benzaldehyde, pubchem1414, 3,5difluorobenzaldehyde, acmc-1aewe, 3,5-diflourobenzaldehyde, 3-5-difluorobenzaldehyde, 3, 5-difluorobenzaldehyde, ksc224o7p, 3,5-difluorophenyl methanone | |
3,5-difluorobenzaldehyde |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
TSCA : TSCA
RUO â Research Use Only