Please login to your online account to display your discounted pricing

3,5-Difluorobenzaldehyde, 98+%, ACROS Organics™


Quantity 2.5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  293470025

 View more versions of this product

Catalog No. AC293470025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Liquid After Melting
Formula Weight 142.1
Quantity 2.5g
Color Clear light green
TSCA TSCA
Density 1.296g/mL
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material 3, 5-Difluorobenzaldehyde
Refractive Index 1.4930 to 1.4950 (20°C, 589nm)
Name Note 98+%
Assay Percent Range 98+%
Molecular Formula C7H4F2O
Linear Formula F2C6H3CHO
MDL Number MFCD00010329
CAS 32085-88-4
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

Specifications

Chemical Name or Material 3, 5-Difluorobenzaldehyde
Name Note 98+%
CAS 32085-88-4
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C7H4F2O
Linear Formula F2C6H3CHO
Formula Weight 142.1
MDL Number MFCD00010329
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

SDS & Safety Documents