missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Difluoroanisole, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 249560050

 View more versions of this product

Catalog No. AC249560050



Specifications

Specifications

3, 5-Difluoroanisole
Authentic
1.4640 to 1.4660 (20°C, 589nm)
5g
93343-10-3
100.0
C7H6F2O
MFCD00042560
OTGQPYSISUUHAF-UHFFFAOYSA-N
1,3-difluoro-5-methoxybenzene
2724518
Liquid
1.23g/mL
Glass Bottle
Colorless
98%
97.5
98%
F2C6H3OCH3
1,3-difluoro-5-methoxy-benzene, 3,5,-difluoroanisole;, 3,5-difluoro anisole, 3,5-difluoro-1-methoxybenzene, 3,5-difluoroanisol, 3,5-difluoroanisole, 3,5-difluoroanisole;, 3.5-difluoroanisole, benzene, 1,3-difluoro-5-methoxy, pubchem1967
COC1=CC(=CC(=C1)F)F
144.121
144.12
≥97.5%

Chemical Identifiers

93343-10-3
144.121
OTGQPYSISUUHAF-UHFFFAOYSA-N
2724518
COC1=CC(=CC(=C1)F)F
C7H6F2O
MFCD00042560
1,3-difluoro-5-methoxy-benzene, 3,5,-difluoroanisole;, 3,5-difluoro anisole, 3,5-difluoro-1-methoxybenzene, 3,5-difluoroanisol, 3,5-difluoroanisole, 3,5-difluoroanisole;, 3.5-difluoroanisole, benzene, 1,3-difluoro-5-methoxy, pubchem1967
1,3-difluoro-5-methoxybenzene
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA

SDS Documents