missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Difluoroanisole, 98%, ACROS Organics™

$128.61 - $128.61

Chemical Identifiers

CAS 93343-10-3
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.121
MDL Number MFCD00042560
InChI Key OTGQPYSISUUHAF-UHFFFAOYSA-N
Synonym 1,3-difluoro-5-methoxy-benzene, 3,5,-difluoroanisole;, 3,5-difluoro anisole, 3,5-difluoro-1-methoxybenzene, 3,5-difluoroanisol, 3,5-difluoroanisole, 3,5-difluoroanisole;, 3.5-difluoroanisole, benzene, 1,3-difluoro-5-methoxy, pubchem1967
PubChem CID 2724518
IUPAC Name 1,3-difluoro-5-methoxybenzene
SMILES COC1=CC(=CC(=C1)F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC249560050
SDS View Documents
Thermo Fisher Scientific
249560050
5g Glass Bottle
Each for $128.61
 
Description

Description

Specifications

Chemical Identifiers

93343-10-3
144.121
OTGQPYSISUUHAF-UHFFFAOYSA-N
2724518
COC1=CC(=CC(=C1)F)F
C7H6F2O
MFCD00042560
1,3-difluoro-5-methoxy-benzene, 3,5,-difluoroanisole;, 3,5-difluoro anisole, 3,5-difluoro-1-methoxybenzene, 3,5-difluoroanisol, 3,5-difluoroanisole, 3,5-difluoroanisole;, 3.5-difluoroanisole, benzene, 1,3-difluoro-5-methoxy, pubchem1967
1,3-difluoro-5-methoxybenzene

Specifications

1.23g/mL
Glass Bottle
Colorless
93343-10-3
100.0
C7H6F2O
MFCD00042560
OTGQPYSISUUHAF-UHFFFAOYSA-N
1,3-difluoro-5-methoxybenzene
2724518
Liquid
3, 5-Difluoroanisole, 98%
Authentic
1.4640 to 1.4660 (20°C, 589nm)
5g
97.5
98%
F2C6H3OCH3
1,3-difluoro-5-methoxy-benzene, 3,5,-difluoroanisole;, 3,5-difluoro anisole, 3,5-difluoro-1-methoxybenzene, 3,5-difluoroanisol, 3,5-difluoroanisole, 3,5-difluoroanisole;, 3.5-difluoroanisole, benzene, 1,3-difluoro-5-methoxy, pubchem1967
COC1=CC(=CC(=C1)F)F
144.121
144.12
≥97.5%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA

SDS