missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Difluoroanisole, 97%

Manufacturer:  Alfa Aesar™ A1549722

 View more versions of this product

Catalog No. AAA1549722



Specifications

Specifications

3,5-Difluoroanisole
145°C to 148°C
93343-10-3
MFCD00042560
4966844
OTGQPYSISUUHAF-UHFFFAOYSA-N
1,3-difluoro-5-methoxybenzene
2724518
97%
43°C (109°F)
100g
C7H6F2O
UN1993
3,5-difluoroanisole, 3,5-difluoro-1-methoxybenzene, benzene, 1,3-difluoro-5-methoxy, 1,3-difluoro-5-methoxy-benzene, 3,5-difluoro anisole, 3,5-difluoroanisol, 3.5-difluoroanisole, pubchem1967, 3,5-difluoroanisole;, 3,5,-difluoroanisole;
COC1=CC(=CC(=C1)F)F
144.121
144.12

Chemical Identifiers

93343-10-3
144.121
OTGQPYSISUUHAF-UHFFFAOYSA-N
2724518
COC1=CC(=CC(=C1)F)F
C7H6F2O
MFCD00042560
3,5-difluoroanisole, 3,5-difluoro-1-methoxybenzene, benzene, 1,3-difluoro-5-methoxy, 1,3-difluoro-5-methoxy-benzene, 3,5-difluoro anisole, 3,5-difluoroanisol, 3.5-difluoroanisole, pubchem1967, 3,5-difluoroanisole;, 3,5,-difluoroanisole;
1,3-difluoro-5-methoxybenzene
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Documents